Order Entry
United Kingdom
ContactUsLinkComponent
Anti-RPP20 Rabbit Polyclonal Antibody
Anti-RPP20 Rabbit Polyclonal Antibody
Catalog # ANTIA10329-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-RPP20 Rabbit Polyclonal Antibody
Catalog # ANTIA10329-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RNaseP protein p20
  • Antigen symbol:
    RPP20
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# O75817
  • Antigen synonyms:
    0610037N12Rik|Ribonuclease P, 20-KD subunit|Ribonucleases P/MRP protein subunit POP7 homolog|POP7 (processing of precursor, S. cerevisiae) homolog|POP7_HUMAN|hPOP7|Ribonuclease P protein subunit p20|processing of precursor 7 ribonuclease P subunit (S. cerevisiae)|processing of precursor 7, S. cerevisiae, homolog of
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
  • Molecular weight:
    14 kDa
  • Sequence:
    MAENREPRGAVEAELDPVEYTLRKRLPSRLPRRPNDIYVNMKTDFKAQLARCQKLLDGGARGQNACSEIYIHGLGLAINRAINIALQLQAGSFGSLQVAANTSTVELVDELEPETDTREPLTRIRNNSAIHIRVFRVTPK
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1 - 140 of human POP7 (NP_005828.2)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to RPP20 for WB with samples derived from human.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: RPP20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human