Anti-CRISPR-Cas9 Rabbit Polyclonal Antibody
New Product
Rabbit polyclonal antibody to CRISPR-Cas9 for WB, ICC/IF and IHC with samples derived from Streptococcus pyogenes.
- Validated applications: WB, ICC/IF, IHC
- Recommended dilutions: WB (on Cas9 transfected cells): 1:1000 to 1:1:2000, WB (on pure Cas9 protein): 1:10000, ICC/IF: 1:1000 to 1:2000
Type: Primary
Antigen: CRISPR-Cas9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Streptococcus pyogenes
- Catalog No:
- ANTIA104346-50
- ANTIA104346-100
- Antigen symbol:
- CRISPR-Cas9
- CRISPR-Cas9
- Antigen name:
- Cas9
- Cas9
- Antigen synonyms:
- CRISPR-associated endonuclease Cas9/Csn1|SpyCas9|CRISPR-Cas9/Csn1|CRISPR-Cas9
- CRISPR-associated endonuclease Cas9/Csn1|SpyCas9|CRISPR-Cas9/Csn1|CRISPR-Cas9
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- S. pyogenes
- S. pyogenes
- Host:
- Rabbit
- Rabbit
- Gene ID:
- UniprotID# Q99ZW2
- UniprotID# Q99ZW2
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant fragment corresponding to amino acids 1 - 608 and amino acids 814 - 1,372 of Streptococcus pyogenes Cas9 (CDJ55032.1), expressed in and purified from E. coli
- Recombinant fragment corresponding to amino acids 1 - 608 and amino acids 814 - 1,372 of Streptococcus pyogenes Cas9 (CDJ55032.1), expressed in and purified from E. coli
- Form:
- Liquid
- Liquid
- Molecular weight:
- 160 kDa
- 160 kDa
- Purification:
- Immunogen affinity purification
- Immunogen affinity purification
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- Supplied in Phosphate Buffered Saline with 50% Glycerol and 5 mM Sodium Azide
- Supplied in Phosphate Buffered Saline with 50% Glycerol and 5 mM Sodium Azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
- Tested applications:
- ICC/IF, IHC
- ICC/IF, IHC
Frequently Bought Together