Anti-NME2 Mouse Monoclonal Antibody [clone: NME2/6433]
Catalog # ANTIA316781-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:C myc purine binding transcription factor PUF
- Antigen symbol:NME2
- Clonality:Monoclonal
- Clone:NME2/6433
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P22392
- Antigen synonyms:epididymis secretory sperm binding protein Li 155an|C-myc purine-binding transcription factor PUF|HEL-S-155an|Histidine protein kinase NDKB|C myc transcription factor|NDKB_HUMAN|NDK B|MGC111212|MGC2212
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:17 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant full-length human NME2 protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [NME2/6433] antibody to NME2 for WB and IHC-P with samples derived from human.
- Validated applications: WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ ml, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: NME2
Clonality: Monoclonal
Clone: NME2/6433
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human