Anti-TLE 1 Rabbit Recombinant Antibody [clone: TLE1/8007R]
Catalog # ANTIA315735-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:E(Sp1) homolog
- Antigen symbol:TLE 1
- Clonality:Recombinant
- Clone:TLE1/8007R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q04724
- Antigen synonyms:GRG1|ESG1|ESG|Enhancer of split groucho-like protein 1|TLE1_HUMAN|Enhancer of split groucho 1|TLE 1|Enhancer of split groucho like protein 1|TLE1
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:83 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human TLE1 fragment (aa 175-338). The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [TLE1/8007R] antibody to TLE 1 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: TLE 1
Clonality: Recombinant
Clone: TLE1/8007R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human