Anti-S100A9 Mouse Monoclonal Antibody [clone: S100A9/7551]
ANTIA315586-100
New Product
- Antibody type:Primary
- Antigen name:60B8AG
- Antigen symbol:S100A9
- Clonality:Monoclonal
- Clone:S100A9/7551
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2 kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P06702
- Antigen synonyms:CAGB|Calprotectin L1H subunit|CFAG|Leukocyte L1 complex heavy chain|L1AG|CGLB|Calgranulin-B|Calgranulin B|Cystic fibrosis antigen B
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:14 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-100) of human S100A9 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [S100A9/7551] antibody to S100A9 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: S100A9
Clonality: Monoclonal
Clone: S100A9/7551
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human