Anti-Melanoma Mouse Recombinant Antibody [clone: rMLANA/8134]
Catalog # ANTIA315295-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Antigen LB39 AA
- Antigen symbol:Melanoma
- Clonality:Recombinant
- Clone:rMLANA/8134
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q16655
- Antigen synonyms:Tumor rejection antigen AB|melan A|Monophenol monooxygenase|Melanoma antigen recognized by T-cells 1|MART1|tyrosinase|Antigen SK29 AA|Melanoma|Melan A protein
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:20 - 22 kDa (doublet)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human MART-1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rMLANA/8134] antibody to melanoma for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Melanoma
Clonality: Recombinant
Clone: rMLANA/8134
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human