Anti-HSP70 Mouse Monoclonal Antibody [clone: HSPA1A/7936]
ANTIA315111-100
New Product
- Antibody type:Primary
- Antigen name:DnaK type molecular chaperone HSP70 1
- Antigen symbol:HSP70
- Clonality:Monoclonal
- Clone:HSPA1A/7936
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P0DMV8.1
- Antigen synonyms:Heat shock 70 kDa protein 1|FLJ54303|FLJ75127|FLJ54408|Epididymis secretory protein Li 103|Heat shock 70 kDa protein 1A/1B|Heat shock 70 kDa protein 1/2|FLJ54370|FLJ54392
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:70 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 200-400) of human HSPA1A protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [HSPA1A/7936] antibody to HSP70 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: HSP70
Clonality: Monoclonal
Clone: HSPA1A/7936
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human