Anti-EpCAM Mouse Monoclonal Antibody [clone: EGP40/1373]
Catalog # ANTIA249247-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Cell surface glycoprotein Trop 2
- Antigen symbol:EpCAM
- Clonality:Monoclonal
- Clone:EGP40/1373
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P16422
- Antigen synonyms:CD326|Antigen identified by monoclonal|Cell surface glycoprotein Trop 1|AUA1|CD326 antigen|Adenocarcinoma-associated antigen|Adenocarcinoma associated antigen|323/A3|17 1A
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:40 - 43 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 77-202, of human EpCAM protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [EGP40/1373] antibody to EpCAM for flow cytometry, WB, IF and IHC-P with samples derived from human.
- Validated applications: Flow cytometry, WB, IF, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, WB: 1 to 2 µg/ml, IF: 1 to 3µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: EpCAM
Clonality: Monoclonal
Clone: EGP40/1373
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human