Anti-Cytokeratin 18 Mouse Monoclonal Antibody [clone: KRT18/1190]
ANTIA249192-100
New Product
- Antibody type:Primary
- Antigen name:Cell proliferation inducing gene 46 protein
- Antigen symbol:Cytokeratin 18
- Clonality:Monoclonal
- Clone:KRT18/1190
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P05783
- Antigen synonyms:CK18|Cell proliferation inducing protein 46|Cytokeratin 18|CYK18|Cytokeratin endo B|Cell proliferation-inducing gene 46 protein|CK 18|CK-18|CYK 18
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:45 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human KRT18 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [KRT18/1190] antibody to Cytokeratin 18 for flow cytometry, IF, WB and IHC-P with samples derived from human and rat.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 18
Clonality: Monoclonal
Clone: KRT18/1190
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat