Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-Puma gamma/GPR109A Rabbit Monoclonal Antibody [clone: ARC54160]
  ANTIA305514-100
New Product
undefined
Anti-Puma gamma/GPR109A Rabbit Monoclonal Antibody [clone: ARC54160]
  ANTIA305514-100
New Product

 

  • Antibody type:
    Primary
  • Antigen symbol:
    Puma gamma / GPR109A
  • Clonality:
    Monoclonal
  • Clone:
    ARC54160
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q8TDS4
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,05% proclin 300
  • Molecular weight:
    42 kDa
  • Sequence:
    CRLMLFMLAMNRQGSIIFLTVVAVDRYFRVVHPHHALNKISNRTAAIISCLLWGITIGLTVHLLKKKMPIQNGGANLCSSFSICHTFQWHEAMFLLEFFLP
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 100 - 200 of human GPR109A/HM74A/HCAR2 (NP_808219.1).
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit monoclonal [ARC54160] antibody to Puma gamma/GPR109A for WB with samples derived from rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:1000 to 1:5000

Type: Primary
Antigen: Puma gamma/GPR109A
Clonality: Monoclonal
Clone: ARC54160
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat