Anti-Neuroligin 3 Mouse Monoclonal Antibody [Clone: S110-29]
Catalog # ANTIA304790-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Gliotactin homolog
- Antigen symbol:Neuroligin 3
- Clonality:Monoclonal
- Clone:S110-29
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q62889
- Antigen synonyms:Nlgn3|NLGN3_HUMAN|HNL3|Neuroligin 3|Neuroligin-3
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,1% sodium azidelak
- Molecular weight:~110 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Fusion protein amino acids 730 - 848 (intracellular C - terminus) of rat Neuroligin - 3. Mouse: 99% identity (118/119 amino acids identical). Human: 98% identity (116/119 amino acids identical) ~60% identity with Neuroligin - 1. ~40% identity with Neuroligin - 2..
- Tested applications:IHC, ICC/IF
- Purification:Protein G purification
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [S110-29] antibody to Neuroligin 3 for WB, IHC and ICC/IF with samples derived from human, mouse and rat.
- Validated applications: WB, IHC, ICC/IF
- Recommended dilutions: WB: 1:1000
Type: Primary
Antigen: Neuroligin 3
Clonality: Monoclonal
Clone: S110-29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat