Anti-Acrolein Mouse Monoclonal Antibody [Clone: 2H2]
ANTIA304754-100
New Product
- Antibody type:Primary
- Antigen name:2-Propen-1-one
- Antigen symbol:Acrolein
- Clonality:Monoclonal
- Clone:2H2
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Species independent
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:Acrylic aldehyde|Acraldehyde|2-propenal|Acrolein
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic Acrolein modified Keyhole Limpet Hemocyanin (KLH)
- Tested applications:ICC/IF, FACS
- Purification:Protein G purification
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [2H2] antibody to Acrolein for WB, ICC/IF, FACS, Flow cytometry and ELISA with samples derived from species independent.
- Validated applications: WB, ICC/IF, FACS, Flow cytometry, ELISA
- Recommended dilutions: WB: 1:1000, ICC/IF: 1:50, ELISA: 1:1000
Type: Primary
Antigen: Acrolein
Clonality: Monoclonal
Clone: 2H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Species Independent