Order Entry
United Kingdom
ContactUsLinkComponent
Mouse SOD2/MnSOD ELISA Kit
Mouse SOD2/MnSOD ELISA Kit
Catalog # ANTIA312792-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Mouse SOD2/MnSOD ELISA Kit
Catalog # ANTIA312792-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Host:
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    SOD2/MnSOD
  • Description:
    Mouse SOD2/MnSOD ELISA kit
  • Environmentally Preferable:
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    3 - 48 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    0,154 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Mouse SOD2/MnSOD ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse SOD2/MnSOD in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 3 to 48 ng/ml
  • Sensitivity: 0,154 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse SOD2/MnSOD ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse SOD2/MnSOD in An antibody specific for SOD2/MnSOD has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SOD2/MnSOD present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-SOD2/MnSOD antibody, which also binds the SOD2/MnSOD present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-SOD2/MnSOD antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-SOD2/MnSOD antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of SOD2/MnSOD captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of SOD2/MnSOD in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.