Specifications
- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Host:
- Primary antibody reactivity:Human
- Target protein:ST14
- Description:Human ST14 ELISA kit
- Environmentally Preferable:
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,75 - 12 ng/ml
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:0,032 ng/ml
- Regulatory status:RUO
Specifications
About this item
Human ST14 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ST14 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 0,75 to 12 ng/ml
- Sensitivity: 0,032 ng/ml
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human ST14 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human ST14 in An antibody specific for ST14 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the ST14 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-ST14 antibody, which also binds the ST14 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-ST14 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-ST14 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of ST14 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of ST14 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.