Specifications
- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Host:
- Primary antibody reactivity:Human
- Target protein:PAEP/Glycodelin
- Description:Human PAEP/Glycodelin ELISA kit
- Environmentally Preferable:
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,75 - 12 µg/ml
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:0,028 µg/ml
- Regulatory status:RUO
Specifications
About this item
Human PAEP/Glycodelin ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PAEP/Glycodelin in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 0,75 to 12 µg/ml
- Sensitivity: 0,028 µg/ml
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human PAEP/Glycodelin ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human PAEP/Glycodelin in serum, plasma or other biological fluids. An antibody specific for PAEP/Glycodelin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PAEP/Glycodelin present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-PAEP/Glycodelin antibody, which also binds the PAEP/Glycodelin present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-PAEP/Glycodelin antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-PAEP/Glycodelin antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of PAEP/Glycodelin captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of PAEP/Glycodelin in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.