Specifications
- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Host:
- Primary antibody reactivity:Human
- Target protein:Histone H1.2
- Description:Human Histone H1.2 ELISA kit
- Environmentally Preferable:
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:150 - 2400 ng/L
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:9,74 ng/L
- Regulatory status:RUO
Specifications
About this item
Human Histone H1.2 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Histone H1.2 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection range: 150 to 2400 ng/L
- Sensitivity: 9,74 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human Histone H1.2 ELISA kit (A313303) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Histone H1.2 in serum, plasma or other biological fluids. An antibody specific for Histone H1.2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Histone H1.2 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Histone H1.2 antibody, which also binds the Histone H1.2 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Histone H1.2 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Histone H1.2 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Histone H1.2 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Histone H1.2 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.