Order Entry
United Kingdom
ContactUsLinkComponent
DH5α Chemically Competent E. coli Cells
DH5α Chemically Competent E. coli Cells
Catalog # ORIGCC100003
Supplier:  OriGene
New Product
DH5α Chemically Competent E. coli Cells
Catalog # ORIGCC100003
Supplier:  OriGene
New Product
Specifications
  • Description:
    DH5α chemically competent cells (≥10⁸ CFU/μg of pUC19 DNA)
  • Environmentally Preferable:
  • Storage conditions:
    The competent cells should be stored at −80 °C, pUC19 control DNA should be stored at −20 °C and SOC recovery medium should be stored at 4 °C immediately upon arrival
  • Genotypes:
    F – φ80lacZDM15 D(lacZYA-argF) U169 recA1 endA1 hsdR17 (rk–, mk+) phoA supE44 l – thi–1 gyrA96 relA1
  • Feature:
    Chemically competent accommodates small and larger plasmids RecA1 and endA1 markers for minimal recombination and enhanced quality of plasmid DNA
  • Pk:
    1 KIT
Specifications
About this item
  • lacZ marker for blue/white colour screening
  • Transformation of cloned DNA into bacterial cells
  • Ideal for subcloning and generating cDNA libraries
Kit contains:
DH5α chemically competent cells (5×200 μl)
Positive control DNA - pUC19 (50 pg/μl, 25 μl)
SOC medium: 20 ml