Order Entry
United Kingdom
ContactUsLinkComponent
IntestiCult™ Organoid Growth Medium (Human)
IntestiCult™ Organoid Growth Medium (Human)
Catalog # STMC06010
CAS Number:  
undefined
IntestiCult™ Organoid Growth Medium (Human)
Catalog # STMC06010
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    IntestiCult™ Organoid growth medium (Human)
  • Environmentally Preferable:
Specifications
About this item
Cell culture medium for establishment and maintenance of human intestinal organoids.
Model key characteristics of the adult intestinal epithelium using intestinal organoids established and maintained with this complete medium formulation and optimised protocol. Using the easy-to-follow and robust protocol, derive organoids from human intestinal crypts in one week; organoid growth across donor samples, including those that are otherwise difficult to grow, is enabled by an enriched stem cell population. Organoids grown in IntestiCult™ Organoid Growth Medium (Human) incorporate a functional lumen enclosed by a polarised intestinal epithelial cell layer and, for versatile modeling applications, can be further differentiated in 3D or in 2D as submerged monolayers or air-liquid interface (ALI) cultures using IntestiCult™ Organoid Differentiation Medium.
Applications of intestinal organoid cultures include studying the development and function of intestinal epithelium, modeling intestinal diseases, and screening molecules for both efficacy and toxicity in an intestinal model. Intestinal organoid cultures can also be used for investigation of adult stem cell properties and for regenerative therapy approaches. Should you intend to use this product for commercial purposes.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').