Order Entry
United Kingdom
ContactUsLinkComponent
Hygromycin B
  STMC03813
 :  
undefined
Hygromycin B
  STMC03813
 :  
  • Pk:
    100 mg
  • Description:
    Hygromycin B
  • Environmentally Preferable:
Antibiotic for selecting transfected prokaryotic and eukaryotic cells.
Hygromycin B, an aminoglycoside antibiotic, inhibits the growth of prokaryotic and eukaryotic cells. Specifically, it inhibits protein synthesis by interfering with translocation of the 70S ribosome subunit and inducing misreading of the mRNA template. Hygromycin B has been used to select transfectants in a wide variety of cells including bacteria, protozoans, yeast, fungi, plants, and mammalian cells. Resistance to Hygromycin B is conferred by a gene coding for a phosphotransferase that phosphorylates Hygromycin B, thereby inactivating it. Hygromycin B is an effective agent for the selection and maintenance of bacterial and eukaryotic cells stably transfected with vectors containing E. coli Hygromycin B resistance gene (hyg or hph). This antibiotic is suitable for use in cell culture for the selection of hygromycin-resistant clones when using ClonaCell™-TCS medium, and for the selection of any hygromycin-resistant transfected cell type.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').