Anti-ERBB2 Recombinant Antibody [Clone: 4D5-8 (trastuzumab)]
Catalog # PRSI24-088
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:erb-b2 receptor tyrosine kinase 2
- Antigen symbol:ERBB2
- Clonality:Recombinant
- Clone:4D5-8 (trastuzumab)
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:2064
- Antigen synonyms:Tyrosine kinase-type cell surface receptor HER2|Metastatic lymph node gene 19 protein (MLN 19)|p185HER2|EGFR|CD340|p185erbB2|Proto-oncogene c-ErbB-2|Proto-oncogene Neu
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:A431 cells (human epidermoid carcinoma) (over)expressing EGFR.
- Size:0,2 mg
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary,Biosimilars
Antigen: erb-b2 receptor tyrosine kinase 2
Clonality: Recombinant
Clone: 4D5-8 (trastuzumab)
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human