- Antibody type:Primary
- Antigen name:Corticotropin-releasing factor receptor 2
- Antigen symbol:CRHR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Format:Purified Antibody
- Form:Liquid
- Gene ID:Q13324
- Antigen synonyms:CRHR2
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at –20 °C. Avoid freez-thaw cycles.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:A recombinant protein fragment from the N-terminal portion of human CRHR2 was used as the immunogen for this antibody (1).
- Purification:Protein A affinity chromatography
- Size:100 µg
- Pk:100 µG
CHCR2, or Corticotropin releasing hormone receptor 2, is a receptor for CRH (corticotropin-releasing hormone), a neurotransmitter secreted by the hypothalamus in response to stress. There are three major isoforms of CHCR2: alpha, beta and gamma, differing in their N-terminus amino acid sequence. Isoform beta and gamma are expressed in the brain, with gamma expression highest in the septum and hippocampus, while alpha is expressed in the peripheral tissues of the brain.
Western blot: 0,5-2 µg/ml Provided assay concentrations are sµggestions only, CRHR2 antibody titration may be required for optimal results.
1. This antibody will detect the alpha (411 AA), beta (438 AA) and gamma (397 AA) isoforms of CRHR2.
Type: Primary
Antigen: CRHR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat