Anti-CBX1 Rabbit Polyclonal Antibody
Catalog # PRSI18-587
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Chromobox homolog 1
- Antigen symbol:CBX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:10951
- Antigen synonyms:HP1Hs-beta|CBX1|p25beta|HP1Hsbeta|HP1-BETA|MOD1|M31|CBX
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-185 of human CBX1 (NP_006798.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CBX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human