Anti-CASC3 Rabbit Polyclonal Antibody
PRSI62-777
New Product
- Antibody type:Primary
- Antigen name:Protein CASC3
- Antigen symbol:CASC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:159-188 aa
- Form:Liquid
- Gene ID:22794
- Modification:None
- Molecular weight:76 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,407 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CASC3 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 159-188 amino acids from human CASC3.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50.
Type: Primary
Antigen: CASC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 159-188
Host: Rabbit
Isotype: Ig
Reactivity: Human