Anti-Pcp2 Rabbit Polyclonal Antibody
Catalog # PRSI62-260
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Purkinje cell protein 2
- Antigen symbol:Pcp2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Mouse
- Environmentally Preferable:
- Epitope:13-41 aa
- Form:Liquid
- Gene ID:18545
- Modification:None
- Molecular weight:13 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This Mouse Pcp2 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 13-41 amino acids from the N-terminal region of mouse Pcp2.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- Pk:400 µl
Specifications
About this item
For IHC-P starting dilution is: 1:500.
Type: Primary
Antigen: Pcp2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 13-41
Host: Rabbit
Isotype: Ig
Reactivity: Mouse