Anti-HIRA Rabbit Polyclonal Antibody
Catalog # PRSI62-213
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Protein HIRA
- Antigen symbol:HIRA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:431-461 aa
- Form:Liquid
- Gene ID:7290
- Modification:None
- Molecular weight:112 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This HIRA antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 431-461 amino acids from the central region of human HIRA.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: HIRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 431-461
Host: Rabbit
Isotype: Ig
Reactivity: Human