Anti-CEL Rabbit Polyclonal Antibody
PRSI23-082
New Product
- Antibody type:Primary
- Antigen name:Carboxyl ester lipase
- Antigen symbol:CEL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:1056
- Antigen synonyms:oncofetal isoform|FAP|BSDL|cholesterol esterase|FAPP|bile salt-dependent lipase|fetoacinar pancreatic protein|CEL|MODY8|LIPA|BAL|carboxyl ester lipase (bile salt-stimulated lipase)|carboxyl ester hydrolase|pancreatic|BSSLL|CEase|bucelipase|bile salt-activated lipase|lysophospholipase|sterol esterase
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide of human CEL
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: CEL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat