Anti-FGFBP1 Rabbit Polyclonal Antibody
PRSI22-955
New Product
- Antibody type:Primary
- Antigen name:Fibroblast growth factor binding protein 1
- Antigen symbol:FGFBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:9982
- Antigen synonyms:FGF-binding protein 1|FGFBP-1|HBP17|17 kDa heparin-binding growth factor-binding protein|FGF-BP|FGF-BP1|FGFBP|17 kDa HBGF-binding protein|heparin-binding growth factor binding protein|fibroblast growth factor-binding protein 1
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 24-234 of human FGFBP1 (NP_005121.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: FGFBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat