Anti-TRIP10 Rabbit Polyclonal Antibody
Catalog # PRSI22-650
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Thyroid hormone receptor interactor 10
- Antigen symbol:TRIP10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:9322
- Antigen synonyms:TRIP10|HSTP|STP|CIP4|TRIP-10|STOT
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 246-545 of human TRIP10 (NP_004231.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: TRIP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse