Anti-CAMK1 Rabbit Polyclonal Antibody
PRSI22-635
New Product
- Antibody type:Primary
- Antigen name:Calcium/calmodulin-dependent protein kinase type 1
- Antigen symbol:CAMK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:8536
- Antigen synonyms:CaM kinase I|CaM kinase I alpha|CaM-KI|CaMKI-alpha|CAMK1|Calcium/calmodulin-dependent protein kinase type 1
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 271-370 of human CAMK1 (NP_003647.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: CAMK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat