Anti-PMS2 Rabbit Polyclonal Antibody
Catalog # PRSI22-577
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:PMS2 postmeiotic segregation increased 2 (S. cerevisiae)
- Antigen symbol:PMS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:5395
- Antigen synonyms:PMS2CL|PMS2|HNPCC4|H_DJ0042M02.9|PMS1 protein homolog 2|PMSL2
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 390-670 of human PMS2 (NP_000526.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PMS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat