Anti-UBE2S Rabbit Polyclonal Antibody
PRSI18-254
New Product
- Antibody type:Primary
- Antigen name:Ubiquitin-conjugating enzyme E2S
- Antigen symbol:UBE2S
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:27338
- Antigen synonyms:E2-EPF|E2EPF|UBE2S|EPF5
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 122-221 of human UBE2S (NP_055316,2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: UBE2S
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat