Anti-PPP1R12B Rabbit Polyclonal Antibody
PRSI16-965
New Product
- Antibody type:Primary
- Antigen name:Protein phosphatase 1 regulatory subunit 12B
- Antigen symbol:PPP1R12B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:4660
- Antigen synonyms:protein phosphatase 1 regulatory subunit 12B|PP1bp55|MYPT2|protein phosphatase 1|myosin phosphatase-targeting subunit 2|regulatory (inhibitor) subunit 12B|myosin phosphatase target subunit 2
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 660-930 of human PPP1R12B (NP_002472,2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: PPP1R12B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human