Anti-KCNQ5 Rabbit Polyclonal Antibody
PRSI16-449
New Product
- Antibody type:Primary
- Antigen name:Potassium voltage-gated channel, KQT-like subfamily, member 5
- Antigen symbol:KCNQ5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:56479
- Antigen synonyms:potassium voltage-gated channel|Kv7.5|OTTHUMP00000064153|OTTHUMP00000064152|KQT-like subfamily|member 5|potassium channel protein|OTTHUMP00000016729|KCNQ5
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 624-923 of human KCNQ5 (NP_001153602,1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: KCNQ5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat