Anti-OPN3 Rabbit Polyclonal Antibody
PRSI16-241
New Product
- Antibody type:Primary
- Antigen name:Opsin 3
- Antigen symbol:OPN3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:23596
- Antigen synonyms:ECPN|ERO|Panopsin|Opsin 3|Opsin-3|NMO-1|OPN3|Encephalopsin
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 300-400 of human OPN3 (NP_055137,2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: OPN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat