Anti-SLC4A4 Rabbit Polyclonal Antibody
PRSI19-594
New Product
- Antibody type:Primary
- Antigen name:Solute carrier family 4 member 4
- Antigen symbol:SLC4A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:8671
- Antigen synonyms:solute carrier family 4 (sodium bicarbonate cotransporter)|kNBC1|brain type|kidney|NBCe1-A|KNBC|member 4|hhNMC|member 5|electrogenic sodium bicarbonate cotransporter 1|NBC2|pancreas)|SLC4A5|HNBC1|sodium bicarbonate cotransporter 1 (sodium bicarbonate cotransporter|pNBC|solute carrier family 4|Na(+)/HCO3(-) cotransporter|NBC1| sodium bicarbonate cotransporter|sodium bicarbonate cotransporter
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human SLC4A4 / NBC (NP_003750.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: SLC4A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat