Anti-MRM3 Rabbit Polyclonal Antibody
Catalog # PRSI19-478
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Mitochondrial rRNA methyltransferase 3
- Antigen symbol:MRM3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:55178
- Antigen synonyms:RNA methyltransferase-like protein 1|16S rRNA [Gm1370] 2'-O-methyltransferase|RNA methyltransferase like 1|mitochondrial|rRNA methyltransferase 3|RNMTL1|putative RNA methyltransferase|16S rRNA (guanosine(1370)-2'-O)-methyltransferase|RMTL1
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-420 of human MRM3 (NP_060616.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: MRM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human