Anti-GNMT Rabbit Polyclonal Antibody
PRSI59-615
New Product
- Antibody type:Primary
- Antigen name:Glycine N-methyltransferase
- Antigen symbol:GNMT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:78 - 106 aa
- Form:Liquid
- Gene ID:27232
- Modification:None
- Molecular weight:33 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This GNMT antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 78 - 106 amino acids from the Central region of human GNMT.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: GNMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 78-106
Host: Rabbit
Isotype: Ig
Reactivity: Human