Anti-VAT1L Rabbit Polyclonal Antibody
Catalog # PRSI56-858
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Synaptic vesicle membrane protein VAT-1 homolog-like
- Antigen symbol:VAT1L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Amino acid range = 391-419
- Form:Liquid
- Gene ID:Q9HCJ6
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:46 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This VAT1L antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 391-419 amino acids from the C-terminal region of human VAT1L
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
KIAA1576 belongs to the zinc-containing alcohol dehydrogenase family; Quinone oxidoreductase subfamily.
- For WB starting dilution is: 1:1000
Type: Primary
Antigen: VAT1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse