Anti-CDRT15L2 Rabbit Polyclonal Antibody
Catalog # PRSI56-491
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:CMT1A duplicated region transcript 15 protein-like protein
- Antigen symbol:CDRT15L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Amino acid range = 134-162
- Form:Liquid
- Gene ID:A8MXV6
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:30 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CDRT15L1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 134-162 amino acids from the Central region of human CDRT15L1
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
The function of this protein remains unknown.
- For WB starting dilution is: 1:1000
Type: Primary
Antigen: CDRT15L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human