Order Entry
United Kingdom
ContactUsLinkComponent
Anti-MEPE Rabbit Polyclonal Antibody
Anti-MEPE Rabbit Polyclonal Antibody
Catalog # PRSI56-396
Supplier:  ProSci Inc.
New Product
undefined
Anti-MEPE Rabbit Polyclonal Antibody
Catalog # PRSI56-396
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Matrix extracellular phosphoglycoprotein
  • Antigen symbol:
    MEPE
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    Amino acid range = 19-48
  • Form:
    Liquid
  • Gene ID:
    Q9NQ76
  • Modification:
    None
  • Storage buffer:
    Supplied in PBS with 0,09% (W/V) sodium azide
  • Molecular weight:
    58 kDa
  • Storage temperature:
    Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    0,33 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This MEPE antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 19-48 amino acids from the N-terminal region of human MEPE
  • Purification:
    This antibody is purified through a protein A column, followed by peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

MEPE seems to play a role in mineralization.

  • For WB starting dilution is: 1:1000

Type: Primary
Antigen: MEPE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human