Anti-ENG Mouse Monoclonal Antibody [clone: STA] (PE (Phycoerythrin))
PRSI76-883
New Product
- Antibody type:Primary
- Antigen name:endoglin
- Antigen symbol:ENG
- Clonality:Monoclonal
- Clone:STA
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:P17813
- Antigen synonyms:ENG
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:100 Tests
The STA monoclonal antibody specifically reacts with human CD106, a 110 kDa glycoprotein also named Vascular Cell Adhesion Molecule-1 (VCAM-1) and INCAM-110. Its expression is induced by inflammation stimuli and cytokines and it is found mainly on activated vascular endothelium. CD106 is also found on subsets of macrophages, stromal cells, dendritic cells, and myeloid lineage cells. The integrins that bind CD106 are CD49d/CD29 and LPAM-1, and it is involved in cell adhesion, transmigration and T cell proliferation.
Type: Primary
Antigen: ENG
Clonality: Monoclonal
Clone: STA
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human