Anti-ZBTB32 Rabbit Polyclonal Antibody
PRSI58-480
New Product
- Antibody type:Primary
- Antigen name:Zinc finger and BTB domain-containing protein 32
- Antigen symbol:ZBTB32
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:96 - 125 aa
- Form:Liquid
- Gene ID:27033
- Modification:None
- Molecular weight:53 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ZBTB32 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 96-125 amino acids from the N-terminal region of human ZBTB32
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: ZBTB32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 96-125
Host: Rabbit
Isotype: Ig
Reactivity: Human