Anti-LAT2 Rabbit Polyclonal Antibody
PRSI58-001
New Product
- Antibody type:Primary
- Antigen name:Linker for activation of T-cells family member 2
- Antigen symbol:LAT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:107 - 135 aa
- Form:Liquid
- Gene ID:7462
- Modification:None
- Molecular weight:27 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This LAT2 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 107-135 amino acids from the Central region of human LAT2
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: LAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 107-135
Host: Rabbit
Isotype: Ig
Reactivity: Human