Anti-TAS1R1 Rabbit Polyclonal Antibody
Catalog # PRSI13-469
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Taste receptor type 1 member 1
- Antigen symbol:TAS1R1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:80835
- Antigen synonyms:Taste receptor type 1 member 1|TAS1R1|TR1|T1R1|GPR70|G-protein coupled receptor 70
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 330-567 of human TAS1R1 (NP_619642.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: TAS1R1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat