Anti-EPPIN Rabbit Polyclonal Antibody
PRSI18-534
New Product
- Antibody type:Primary
- Antigen name:Epididymal peptidase inhibitor
- Antigen symbol:EPPIN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:57119
- Antigen synonyms:epididymis secretory sperm binding protein|CT72|SPINLW1|eppin|cancer/testis antigen 72|CT71|epididymal protease inhibitor|cancer/testis antigen 71|WAP four-disulfide core domain protein 7|WAP7|with Kunitz and WAP domains 1 (eppin)|WFDC7|dJ461P17.2|protease inhibitor WAP7|serine peptidase inhibitor-like
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 22-133 of human EPPIN (NP_065131.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: EPPIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse