Anti-NDRG1 Rabbit Polyclonal Antibody
PRSI18-529
New Product
- Antibody type:Primary
- Antigen name:N-myc downstream regulated 1
- Antigen symbol:NDRG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:10397
- Antigen synonyms:TDD5|RIT42|HMSNL|PROXY1|DRG-1|GC4|NDR1|NDRG1|CAP43|TARG1|NMSL|CMT4D|RTP|DRG1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 175-394 of human NDRG1 (NP_001128714.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: NDRG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat