Anti-DDX53 Rabbit Polyclonal Antibody
PRSI62-946
New Product
- Antibody type:Primary
- Antigen name:Probable ATP-dependent RNA helicase DDX53
- Antigen symbol:DDX53
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:597-626 aa
- Form:Liquid
- Gene ID:168400
- Modification:None
- Molecular weight:71 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This DDX53 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 597-626 amino acids from the C-terminal region of human DDX53.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:400 µl
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: DDX53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 597-626
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse