Anti-DEPDC7 Rabbit Polyclonal Antibody
PRSI60-853
New Product
- Antibody type:Primary
- Antigen name:DEP domain-containing protein 7
- Antigen symbol:DEPDC7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:439 - 467 aa
- Form:Liquid
- Gene ID:91614
- Modification:None
- Molecular weight:58 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,31 mg/ml
- Shipping temperature:4 °C
- Immunogen:This DEPDC7 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 439 - 467 amino acids from the C-terminal region of human DEPDC7.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: DEPDC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 439-467
Host: Rabbit
Isotype: Ig
Reactivity: Human