Anti-GLOD4 Rabbit Polyclonal Antibody
Vial
Catalog # PRSI23-814
Supplier: ProSci Inc.
Jump to:
Product Details & Documents
Type: Primary
Antigen: GLOD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Pack type:Vial
- Western blot:Yes
- Clonality:Polyclonal
- Modification:None
- Purification:Affinity purification.
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Host:Rabbit
- Concentration:Batch dependent
- Shipping temperature:20
- Antigen name:glyoxalase domain containing 4
- Cat. No.:PRSI23-814
- Gene ID:51031
- Antigen synonyms:glyoxalase domain-containing protein 4|C17orf25|CGI-15HC71
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 179-298 of human GLOD4 (NP_057164.3)
- Conjugation:Unconjugated
- Antigen symbol:GLOD4
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



