Anti-MGEA5 Rabbit Polyclonal Antibody
PRSI23-776
New Product
- Antibody type:Primary
- Antigen name:O-GlcNAcase
- Antigen symbol:MGEA5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:10724
- Antigen synonyms:N-acetyl-beta-glucosaminidase|MEA5|beta-hexosaminidase|hyaluronidase in meningioma|nuclear cytoplasmic O-GlcNAcase and acetyltransferase|N-acetyl-beta-D-glucosaminidase|bifunctional protein NCOAT|meningioma-expressed antigen 5|MGEA5|beta-N-acetylhexosaminidase|beta-N-acetylglucosaminidase|NCOAT|meningioma expressed antigen 5 (hyaluronidase)|protein O-GlcNAcase
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 380-580 of human MGEA5 (NP_001135906.1)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: MGEA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat